Learn More
Apexbio Technology LLC Olmesartan 144689-24-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A36815.1
Encompass_Preferred
Olmesartan (CAS 144689-24-7) is a small molecule angiotensin II receptor antagonist primarily acting by selectively blocking the binding of angiotensin II to AT1 receptors This mechanism interrupts angiotensin II-mediated vasoconstriction and aldosterone secretion resulting in antihypertensive effects In biomedical research Olmesartan serves as a valuable tool for examining hypertensive disease models elucidating angiotensin II signaling pathways and exploring therapeutic strategies targeting cardiovascular regulation
Catalog No. 50-000-64158