Learn More
TARGETMOL CHEMICALS INC OLMESARTAN 500MG
SDP
Catalog No. 502579725
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olmesartan (RNH 6270) is an Angiotensin II Type I receptor antagonist. Olmesartan is the active form of the antihypertensive drug olmesartan medoxomiland has antihypertensive activity. purity: 99%
Catalog No. 50-257-9725 Supplier TARGETMOL CHEMICALS INC Supplier No. T5964500MG
Also available in 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olmesartan (RNH 6270) is an Angiotensin II Type I receptor antagonist. Olmesartan is the active form of the antihypertensive drug olmesartan medoxomiland has antihypertensive activity. purity: 99%