Learn More
Medchemexpress LLC Olmesartan-d4 | 1420880-41-6 | 98.8% | 450.53 | 5 MG
SDP
Catalog No. 5000391251
Encompass_Preferred
Olmesartan-d4 (RNH-6270-d4) is the deuterium-labeled version of Olmesartan, an angiotensin II receptor (AT1R) antagonist. It is primarily used to treat high blood pressure. This compound can serve as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitation during drug development
  • Can be an internal standard for quantitative analysis
  • AT1 receptor antagonist
  • Deuterium-labeled compound
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-003-91251 Supplier Medchemexpress LLC Supplier No. HY17004S5MG
Olmesartan-d4 (RNH-6270-d4) is the deuterium-labeled version of Olmesartan, an angiotensin II receptor (AT1R) antagonist. It is primarily used to treat high blood pressure. This compound can serve as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitation during drug development
  • Can be an internal standard for quantitative analysis
  • AT1 receptor antagonist
  • Deuterium-labeled compound
  • Suitable for NMR, GC-MS, or LC-MS applications