Learn More
Medchemexpress LLC Olmesartan-d4 (RNH-6270-d4) | 1420880-41-6 | 98.8% | 450.53 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17004S1MG
Encompass_Preferred
Olmesartan-d4 is a deuterium-labeled form of Olmesartan, an angiotensin II receptor (AT1R) antagonist primarily used for treating high blood pressure.
  • Functions as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.
  • Angiotensin II receptor (AT1R) antagonist.
  • Aids in the treatment of high blood pressure.
Catalog No. 50-003-91250