Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olmesartan-d4 (RNH-6270-d4) | 1420880-41-6 | MFCD10567132 | 98.8% | 450.53 | C24H22D4N6O3 | 10 MG
SDP

Catalog No. 5000295177
Encompass_Preferred

Olmesartan-d4 is a deuterium-labeled analogue of olmesartan supplied as a 10 mg research-grade standard for laboratory analytical use. It is intended as a tracer or internal standard for quantitative methods and is not for human or veterinary use.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 98.83% suitable for analytical assays.
  • Molecular weight 450.53; formula C24H22D4N6O3.
  • Soluble in DMSO (20.83 mg/mL); may require ultrasonic treatment.
  • Recommended storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).
  • Supplied as a 10 mg quantity for analytical workflows.

Catalog No. 50-002-95177 Supplier Medchemexpress LLC Supplier No. HY17004S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olmesartan-d4 is a deuterium-labeled analogue of olmesartan supplied as a 10 mg research-grade standard for laboratory analytical use. It is intended as a tracer or internal standard for quantitative methods and is not for human or veterinary use.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 98.83% suitable for analytical assays.
  • Molecular weight 450.53; formula C24H22D4N6O3.
  • Soluble in DMSO (20.83 mg/mL); may require ultrasonic treatment.
  • Recommended storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).
  • Supplied as a 10 mg quantity for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.