Learn More
Medchemexpress LLC Olmesartan lactone impurity | 849206-43-5 | MFCD27978125 | 99.3% | 428.49 g/mol | C24H24N6O2 | 5 MG
SDP
Catalog No. 5000262561
Encompass_Preferred
Olmesartan lactone impurity is the cyclic ester impurity of olmesartan, provided as a high-purity analytical standard for research, impurity profiling, and method development. The compound has molecular formula C24H24N6O2 and molecular weight 428.49 g/mol, and is supplied in small-mass quantities for laboratory use.
  • Reported purity 99.27% (rounded to 99.3%).
  • Molecular formula C24H24N6O2; molecular weight 428.49 g/mol.
  • Suitable as a reference standard for impurity profiling and analytical method development.
  • Available in small pack sizes for analytical testing, such as 5 mg and 10 mg.
  • Analytical documents available: data sheet, COA, HNMR, LCMS, and SDS.
Catalog No. 50-002-62561 Supplier Medchemexpress LLC Supplier No. HY1312765MG
Olmesartan lactone impurity is the cyclic ester impurity of olmesartan, provided as a high-purity analytical standard for research, impurity profiling, and method development. The compound has molecular formula C24H24N6O2 and molecular weight 428.49 g/mol, and is supplied in small-mass quantities for laboratory use.
  • Reported purity 99.27% (rounded to 99.3%).
  • Molecular formula C24H24N6O2; molecular weight 428.49 g/mol.
  • Suitable as a reference standard for impurity profiling and analytical method development.
  • Available in small pack sizes for analytical testing, such as 5 mg and 10 mg.
  • Analytical documents available: data sheet, COA, HNMR, LCMS, and SDS.