Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olmesartan lactone impurity | 849206-43-5 | MFCD27978125 | 99.3% | C24H24N6O2 | 10MG
SDP

Catalog No. 5000259886
Encompass_Preferred

Olmesartan lactone impurity is a cyclic ester impurity of olmesartan supplied as an analytical reference standard for impurity profiling, stability studies, and analytical method development. It is a white to off-white solid with high reported purity and specified storage conditions to support long-term stability.

  • High purity: 99.27%.
  • Molecular formula C24H24N6O2 and molecular weight 428.49 g·mol⁻¹.
  • Available in small, measured quantities suitable for analytical use.
  • Storage conditions provided for powder and solution to maintain integrity.
  • Intended for impurity profiling, stability testing, and method validation.

Catalog No. 50-002-59886 Supplier Medchemexpress LLC Supplier No. HY13127610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olmesartan lactone impurity is a cyclic ester impurity of olmesartan supplied as an analytical reference standard for impurity profiling, stability studies, and analytical method development. It is a white to off-white solid with high reported purity and specified storage conditions to support long-term stability.

  • High purity: 99.27%.
  • Molecular formula C24H24N6O2 and molecular weight 428.49 g·mol⁻¹.
  • Available in small, measured quantities suitable for analytical use.
  • Storage conditions provided for powder and solution to maintain integrity.
  • Intended for impurity profiling, stability testing, and method validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.