Learn More
TARGETMOL CHEMICALS INC Olmesartan Medoxomil 200MG
SDP
Catalog No. 502876250
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Olmesartan Medoxomil (Benicar) is an angiotensin II type 1 receptor blocker that is used to manage hypertension. Purity 98.92%
Catalog No. 50-287-6250 Supplier TARGETMOL CHEMICALS INC Supplier No. T1518200MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Olmesartan Medoxomil (Benicar) is an angiotensin II type 1 receptor blocker that is used to manage hypertension. Purity 98.92%