Learn More
Cayman Chemical Olmesartn MedoxomIl 50mg
SDP
Catalog No. 502952213
Encompass_Preferred
A prodrug form of olmesartan; inhibits the angiotensin II pressor response in normotensive rats at 0.1 mg/kg; reduces both systolic and diastolic blood pressure in fructose-fed rats, a model of insulin resistance, at 10 mg/kg per day, as well as in chow-fed control animals at 11.4 mg/kg per day; inhibits increases in plasma levels of triglycerides and non-esterified fatty acids in fructose-fed rats
Catalog No. 50-295-2213 Supplier Cayman Chemical Supplier No. 1161450 MG
A prodrug form of olmesartan; inhibits the angiotensin II pressor response in normotensive rats at 0.1 mg/kg; reduces both systolic and diastolic blood pressure in fructose-fed rats, a model of insulin resistance, at 10 mg/kg per day, as well as in chow-fed control animals at 11.4 mg/kg per day; inhibits increases in plasma levels of triglycerides and non-esterified fatty acids in fructose-fed rats