Learn More
Medchemexpress LLC Olmutinib | 1353550-13-6 | 99.5% / 99.9% | 486.59 | 200 MG
SDP
Catalog No. 5000400941
Encompass_Preferred
Olmutinib is an orally active and irreversible third-generation EGFR tyrosine kinase inhibitor. It binds to a cysteine residue near the kinase domain and is used for Non-Small Cell Lung Cancer (NSCLC). This product is for research use only.
  • Orally active
  • Irreversible third-generation EGFR tyrosine kinase inhibitor
  • Binds to a cysteine residue near the kinase domain
  • Used for Non-Small Cell Lung Cancer (NSCLC)
  • For research use only
Catalog No. 50-004-00941 Supplier Medchemexpress LLC Supplier No. HY19730200MG
Olmutinib is an orally active and irreversible third-generation EGFR tyrosine kinase inhibitor. It binds to a cysteine residue near the kinase domain and is used for Non-Small Cell Lung Cancer (NSCLC). This product is for research use only.
  • Orally active
  • Irreversible third-generation EGFR tyrosine kinase inhibitor
  • Binds to a cysteine residue near the kinase domain
  • Used for Non-Small Cell Lung Cancer (NSCLC)
  • For research use only