Learn More
Medchemexpress LLC Olmutinib | 1353550-13-6 | 99.5% | 486.59 | 5 MG
SDP
Catalog No. 501873762
Encompass_Preferred
Olmutinib is an orally active and irreversible third EGFR tyrosine kinase inhibitor that binds to a cysteine residue near the kinase domain. It is used for non-small cell lung cancer.
  • Orally active.
  • Irreversible third EGFR tyrosine kinase inhibitor.
  • Binds to a cysteine residue near the kinase domain.
  • Used for non-small cell lung cancer.
Catalog No. 50-187-3762 Supplier Medchemexpress LLC Supplier No. HY197305MG
Olmutinib is an orally active and irreversible third EGFR tyrosine kinase inhibitor that binds to a cysteine residue near the kinase domain. It is used for non-small cell lung cancer.
  • Orally active.
  • Irreversible third EGFR tyrosine kinase inhibitor.
  • Binds to a cysteine residue near the kinase domain.
  • Used for non-small cell lung cancer.