Learn More
Medchemexpress LLC HY-13106 50mg , Olodanrigan CAS:1316755-16-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310650MG
Encompass_Preferred
HY-13106 50mg Olodanrigan CAS: 1316755-16-4 Olodanrigan (EMA401) is a highly selective, orally active, peripherally restricted angiotensin II type 2 receptor (AT2R) antagonist. It is under development as a neuropathic pain therapeutic agent. Olodanrigan (EMA401) analgesic action appears to involve inhibition of augmented AngII/AT2R induced p38 and p42/p44 MAPK activation, and hence inhibition of DRG neuron hyperexcitability and sprouting of DRG neurons[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0038