Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olodaterol | 868049-49-4 | MFCD16619349 | 98.7% | C21H26N2O5 | 10MG
SDP

Catalog No. 5000261400
Encompass_Preferred

Olodaterol is a selective, long-acting β2-adrenoceptor agonist supplied as a research reagent for studies of bronchodilation and β2-adrenergic signaling. The compound is characterized by its molecular formula and analytical documentation, and is provided with handling and storage guidance to maintain stability.

  • Selective long-acting β2-adrenoceptor agonist for receptor pharmacology studies.
  • Supplied as a characterized research reagent with certificate of analysis.
  • High purity suitable for research (COA-reported batch purity ~98.7%).
  • Molecular formula C21H26N2O5 and molecular weight 386.44 g/mol.
  • Storage and handling recommendations included to preserve compound stability.
  • Suitable for biochemical, cellular, and analytical applications.

Catalog No. 50-002-61400 Supplier Medchemexpress LLC Supplier No. HY1430110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olodaterol is a selective, long-acting β2-adrenoceptor agonist supplied as a research reagent for studies of bronchodilation and β2-adrenergic signaling. The compound is characterized by its molecular formula and analytical documentation, and is provided with handling and storage guidance to maintain stability.

  • Selective long-acting β2-adrenoceptor agonist for receptor pharmacology studies.
  • Supplied as a characterized research reagent with certificate of analysis.
  • High purity suitable for research (COA-reported batch purity ~98.7%).
  • Molecular formula C21H26N2O5 and molecular weight 386.44 g/mol.
  • Storage and handling recommendations included to preserve compound stability.
  • Suitable for biochemical, cellular, and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.