Learn More
Medchemexpress LLC Olodaterol-d3 hydrochloride | 425.92 | C21H24D3ClN2O5 | 1mg
SDP
Catalog No. 5000269374
Encompass_Preferred
Olodaterol-d3 hydrochloride (BI1744-d3 hydrochloride) is the deuterium labeled Olodaterol hydrochloride (HY-14301A) Olodaterol (BI1744) hydrochloride is a selective long acting 2-adrenoceptor ( 2-AR) agonist (EC50 0 1 nM and pKi 9 14 for human 2-adrenoceptor respectively) Olodaterol can be used for chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis[1][2][3]
Catalog No. 50-002-69374 Supplier Medchemexpress LLC Supplier No. HYW7446251MG
Olodaterol-d3 hydrochloride (BI1744-d3 hydrochloride) is the deuterium labeled Olodaterol hydrochloride (HY-14301A) Olodaterol (BI1744) hydrochloride is a selective long acting 2-adrenoceptor ( 2-AR) agonist (EC50 0 1 nM and pKi 9 14 for human 2-adrenoceptor respectively) Olodaterol can be used for chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis[1][2][3]