Learn More
Medchemexpress LLC Olodaterol hydrochloride | 869477-96-3 | >99.3% | C₂₁H₂₇ClN₂O₅ | 1 MG
SDP
Catalog No. 5000353836
Encompass_Preferred
Olodaterol hydrochloride is a high-purity chemical compound primarily utilized for research and analytical studies. This compound functions as a β2-adrenergic receptor agonist and is often investigated for its potential applications in conditions such as chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis. It is provided as a standard for various analytical purposes.
  • Used for research and analytical studies
  • Functions as a β2-adrenergic receptor agonist
  • High purity verified by HPLC
  • Defined chemical formula C₂₁H₂₇ClN₂O₅
  • Potential relevance in chronic obstructive pulmonary disease and pulmonary fibrosis research
Catalog No. 50-003-53836 Supplier Medchemexpress LLC Supplier No. HY14301AR1MG
Olodaterol hydrochloride is a high-purity chemical compound primarily utilized for research and analytical studies. This compound functions as a β2-adrenergic receptor agonist and is often investigated for its potential applications in conditions such as chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis. It is provided as a standard for various analytical purposes.
  • Used for research and analytical studies
  • Functions as a β2-adrenergic receptor agonist
  • High purity verified by HPLC
  • Defined chemical formula C₂₁H₂₇ClN₂O₅
  • Potential relevance in chronic obstructive pulmonary disease and pulmonary fibrosis research