Learn More
Medchemexpress LLC Olomoucine | 101622-51-9 | 298.34 | 5 MG
SDP
Catalog No. 5000454990
Encompass_Preferred
Olomoucine is an ATP competitive inhibitor of CDKs. It is a purine derivative that regulates the cell cycle and demonstrates anti-melanin tumor activity.
  • Inhibits CDC2/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E (both IC50=7 μM)
  • Inhibits CDK/p35 kinase (IC50=3 μM)
  • Inhibits ERK1/p44 MAP kinase (IC50=25 μM)
  • Induces apoptosis in tumor cells
  • Target: CDK
  • Pathway: Cell Cycle/DNA damage
Catalog No. 50-004-54990 Supplier Medchemexpress LLC Supplier No. HYW0114285MG
Olomoucine is an ATP competitive inhibitor of CDKs. It is a purine derivative that regulates the cell cycle and demonstrates anti-melanin tumor activity.
  • Inhibits CDC2/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E (both IC50=7 μM)
  • Inhibits CDK/p35 kinase (IC50=3 μM)
  • Inhibits ERK1/p44 MAP kinase (IC50=25 μM)
  • Induces apoptosis in tumor cells
  • Target: CDK
  • Pathway: Cell Cycle/DNA damage