Learn More
TARGETMOL CHEMICALS INC OLOMOUCINE 5MG
SDP
Catalog No. 502581019
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olomoucine is an ATP competitive inhibitor of Cdk2/cyclin A Cdc2/CyclinB CDK2/CyclinE CDK5/p35 and ERK1/p44 MAP kinase with IC50s of 7 7 7 3 and 25 µM respectively. Olomoucine regulates cell cycle and exhibits anti-melanin tumor activities. purity: 99%
Catalog No. 50-258-1019 Supplier TARGETMOL CHEMICALS INC Supplier No. T215885MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olomoucine is an ATP competitive inhibitor of Cdk2/cyclin A Cdc2/CyclinB CDK2/CyclinE CDK5/p35 and ERK1/p44 MAP kinase with IC50s of 7 7 7 3 and 25 µM respectively. Olomoucine regulates cell cycle and exhibits anti-melanin tumor activities. purity: 99%