Learn More
Abcam Olomoucine, ATP-competitive CDK inhibitor, 25MG
Catalog No. 502707109
Encompass_Preferred
MW 298.35 g/mol, Purity >98%. ATP-competitive CDK inhibitor (IC₅₀ values are 7 (CDC2/cyclin B), 7 (CDK2/cyclin A), 7 (CDK2/cyclin E), 3 (CDK/p35 kinase) and 25 μM (ERK1/p44 MAPK)). Inhibits cell cycle progression in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7109 Supplier Abcam Supplier No. AB12093825MG
MW 298.35 g/mol, Purity >98%. ATP-competitive CDK inhibitor (IC₅₀ values are 7 (CDC2/cyclin B), 7 (CDK2/cyclin A), 7 (CDK2/cyclin E), 3 (CDK/p35 kinase) and 25 μM (ERK1/p44 MAPK)). Inhibits cell cycle progression in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement