Learn More
Selleck Chemical LLC Olopatadine HCl S2494-10mg
SDP
Catalog No. 503112149
Encompass_Preferred
Olopatadine HCl(ALO4943A KW4679) is a histamine blocker and mast cell stabilizer with an IC50 of 559 M for the release of histamine
Catalog No. 50-311-2149 Supplier Selleck Chemical LLC Supplier No. S249410MG
Add to Cart
Add to Cart
Olopatadine HCl(ALO4943A KW4679) is a histamine blocker and mast cell stabilizer with an IC50 of 559 M for the release of histamine