Learn More
Apexbio Technology LLC Olopatadine HCl 140462-76-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090611
Encompass_Preferred
Olopatadine HCl (CAS 140462-76-6) is a small molecule antagonist of the histamine H1 receptor classified as a second-generation antihistamine By selectively blocking H1 receptors olopatadine inhibits histamine-mediated inflammatory responses in target tissues Preclinical studies have shown that olopatadine suppresses allergic manifestations such as ear swelling indicating its potential to modulate acute hypersensitivity reactions This compound is widely utilized in research focused on the mechanisms of allergic conjunctivitis and other histamine-dependent allergic disorders
Catalog No. 50-000-90611 Supplier Apexbio Technology LLC Supplier No. B15765.1
Olopatadine HCl (CAS 140462-76-6) is a small molecule antagonist of the histamine H1 receptor classified as a second-generation antihistamine By selectively blocking H1 receptors olopatadine inhibits histamine-mediated inflammatory responses in target tissues Preclinical studies have shown that olopatadine suppresses allergic manifestations such as ear swelling indicating its potential to modulate acute hypersensitivity reactions This compound is widely utilized in research focused on the mechanisms of allergic conjunctivitis and other histamine-dependent allergic disorders