Learn More
TARGETMOL CHEMICALS INC OLOPATADINE HYDROCHLORID 100MG
SDP
Catalog No. 502577447
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olopatadine hydrochloride (ALO4943A) is an antihistamine with mast-cell stabilizing properties used as eye drops in the treatment of ALLERGIC CONJUNCTIVITIS. purity: 99%
Catalog No. 50-257-7447 Supplier TARGETMOL CHEMICALS INC Supplier No. T1170100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olopatadine hydrochloride (ALO4943A) is an antihistamine with mast-cell stabilizing properties used as eye drops in the treatment of ALLERGIC CONJUNCTIVITIS. purity: 99%