Learn More
Medchemexpress LLC Olopatadine hydrochloride | 140462-76-6 | 99.8% | 100 MG
SDP
Catalog No. 5000408910
Encompass_Preferred
Olopatadine hydrochloride (ALO4943A) is a histamine blocker primarily used in the treatment of allergic conjunctivitis. As a second-generation histamine H1 receptor antagonist, it also demonstrates effectiveness for allergic rhinitis and chronic urticaria. It is characterized by rapid and high absorption, with significant renal clearance in humans.
  • Significantly inhibits ear swelling and increased production of IL-4, IL-1beta, IL-6, GM-CSF, and NGF in lesioned ear tissue.
  • Inhibits histamine release from human conjunctival mast cell preparations in vitro.
  • Attenuates passive anaphylaxis in guinea pig conjunctiva.
Catalog No. 50-004-08910 Supplier Medchemexpress LLC Supplier No. HYB0426A100MG
Olopatadine hydrochloride (ALO4943A) is a histamine blocker primarily used in the treatment of allergic conjunctivitis. As a second-generation histamine H1 receptor antagonist, it also demonstrates effectiveness for allergic rhinitis and chronic urticaria. It is characterized by rapid and high absorption, with significant renal clearance in humans.
  • Significantly inhibits ear swelling and increased production of IL-4, IL-1beta, IL-6, GM-CSF, and NGF in lesioned ear tissue.
  • Inhibits histamine release from human conjunctival mast cell preparations in vitro.
  • Attenuates passive anaphylaxis in guinea pig conjunctiva.