Learn More
Medchemexpress LLC Olopatadine hydrochloride | 5 MG
SDP
Catalog No. 5000406534
Encompass_Preferred
Olopatadine hydrochloride is a second-generation selective histamine H1-receptor antagonist and a potent anti-allergic drug with mast cell stabilizing and anti-histamine properties. It is primarily used for the treatment of allergic conjunctivitis and allergic rhinitis. It significantly inhibits ear swelling and the production of several interleukins (IL-4, IL-1beta, IL-6), GM-CSF, and NGF in lesioned ears. It is rapidly absorbed and inhibits histamine release from human conjunctival mast cells in vitro.
  • Second-generation selective histamine H1-receptor antagonist
  • Potent anti-allergic drug
  • Mast cell stabilizing and anti-histamine properties
  • Used to treat allergic conjunctivitis and allergic rhinitis
  • Inhibits ear swelling and production of IL-4, IL-1beta, IL-6, GM-CSF, and NGF
  • Rapidly absorbed in healthy human volunteers
  • Inhibits histamine release from human conjunctival mast cell preparations in vitro
Catalog No. 50-004-06534 Supplier Medchemexpress LLC Supplier No. HYB0426A5MG
Olopatadine hydrochloride is a second-generation selective histamine H1-receptor antagonist and a potent anti-allergic drug with mast cell stabilizing and anti-histamine properties. It is primarily used for the treatment of allergic conjunctivitis and allergic rhinitis. It significantly inhibits ear swelling and the production of several interleukins (IL-4, IL-1beta, IL-6), GM-CSF, and NGF in lesioned ears. It is rapidly absorbed and inhibits histamine release from human conjunctival mast cells in vitro.
  • Second-generation selective histamine H1-receptor antagonist
  • Potent anti-allergic drug
  • Mast cell stabilizing and anti-histamine properties
  • Used to treat allergic conjunctivitis and allergic rhinitis
  • Inhibits ear swelling and production of IL-4, IL-1beta, IL-6, GM-CSF, and NGF
  • Rapidly absorbed in healthy human volunteers
  • Inhibits histamine release from human conjunctival mast cell preparations in vitro