Learn More
Cayman Chemical OlopatdInhydrochlorIde 10mg
SDP
Catalog No. 502955789
Encompass_Preferred
A histamine H1 receptor antagonist (Ki =41 nM); 1,059- and 4,177-fold selective for histamine H1 over H2 and H3 receptors, respectively; inhibits histamine-induced phosphoinositide turnover in isolated human conjunctival epithelial cells, isolated human corneal fibroblasts, and human TM3 cells (IC50s = 9.5, 19, and 39.9 nM, respectively); inhibits passive cutaneous anaphylaxis in rats (ED50 = 49 UG/kg) and IgG1-mediated bronchoconstriction in ovalbumin-sensitized guinea pigs (ED50 = 30 UG/kg)
Catalog No. 50-295-5789 Supplier Cayman Chemical Supplier No. 1199910 MG
A histamine H1 receptor antagonist (Ki =41 nM); 1,059- and 4,177-fold selective for histamine H1 over H2 and H3 receptors, respectively; inhibits histamine-induced phosphoinositide turnover in isolated human conjunctival epithelial cells, isolated human corneal fibroblasts, and human TM3 cells (IC50s = 9.5, 19, and 39.9 nM, respectively); inhibits passive cutaneous anaphylaxis in rats (ED50 = 49 UG/kg) and IgG1-mediated bronchoconstriction in ovalbumin-sensitized guinea pigs (ED50 = 30 UG/kg)