Learn More
Cayman Chemical Olopatadine (hydrochloride), Purity: ≥98%, CAS: 140462-76-6, 50 mg
SDP
Catalog No. 502953936
Encompass_Preferred
A histamine H1 receptor antagonist (Ki =41 nM); 1,059- and 4,177-fold selective for histamine H1 over H2 and H3 receptors, respectively; inhibits histamine-induced phosphoinositide turnover in isolated human conjunctival epithelial cells, isolated human corneal fibroblasts, and human TM3 cells (IC50s = 9.5, 19, and 39.9 nM, respectively); inhibits passive cutaneous anaphylaxis in rats (ED50 = 49 µg/kg) and IgG1-mediated bronchoconstriction in ovalbumin-sensitized guinea pigs (ED50 = 30 µg/kg)
Catalog No. 50-295-3936 Supplier Cayman Chemical Supplier No. 1199950 MG
A histamine H1 receptor antagonist (Ki =41 nM); 1,059- and 4,177-fold selective for histamine H1 over H2 and H3 receptors, respectively; inhibits histamine-induced phosphoinositide turnover in isolated human conjunctival epithelial cells, isolated human corneal fibroblasts, and human TM3 cells (IC50s = 9.5, 19, and 39.9 nM, respectively); inhibits passive cutaneous anaphylaxis in rats (ED50 = 49 µg/kg) and IgG1-mediated bronchoconstriction in ovalbumin-sensitized guinea pigs (ED50 = 30 µg/kg)