Learn More
Cayman Chemical oloyl EthnolamID phsphate 10mg
SDP
Catalog No. 502995594
Encompass_Preferred
An LPA mimetic, LPA receptor agonist, and phosphate ester of OEA; selectively increases [35S]GTPγS binding to HEK293T cell membranes expressing LPA1 and LPA2 over LPA3 receptors; induces calcium mobilization and inhibits forskolin-induced cAMP accumulation in MDA-MB-231 cells (EC50s = 1.2 and 101 nM, respectively)
Catalog No. 50-299-5594 Supplier Cayman Chemical Supplier No. 2288610 MG
Add to Cart
Add to Cart
An LPA mimetic, LPA receptor agonist, and phosphate ester of OEA; selectively increases [35S]GTPγS binding to HEK293T cell membranes expressing LPA1 and LPA2 over LPA3 receptors; induces calcium mobilization and inhibits forskolin-induced cAMP accumulation in MDA-MB-231 cells (EC50s = 1.2 and 101 nM, respectively)