Learn More
Medchemexpress LLC Olprinone hydrochloride | 119615-63-3 | 99.9% | C14H11ClN4O | 25 MG
SDP
Catalog No. 5000356812
Encompass_Preferred
Olprinone Hydrochloride, also known as Loprinone Hydrochloride, is a potent phosphodiesterase (PDE) 3 inhibitor with an IC50 of 0.35 μM. It is utilized in research for heart failure due to its positive inotropic and vasodilative effects, and it also demonstrates anti-inflammatory activity.
  • Potent PDE3 inhibitor (IC50 of 0.35 μM)
  • Used in research for heart failure
  • Exhibits positive inotropic and vasodilative effects
  • Demonstrates anti-inflammatory activity
  • Modulates inflammation associated with myocardial ischemia-reperfusion injury in rats
  • Reduces histological evidence of myocardial injury
  • Decreases pro-inflammatory cytokines (TNF-α and IL-1β)
  • Reduces adhesion molecules (ICAM-1 and P-Selectin)
  • Decreases nitrotyrosine formation and NF-κB expression
  • Reduces PAR formation and apoptosis
Catalog No. 50-003-56812 Supplier Medchemexpress LLC Supplier No. 25MGHY14254
Olprinone Hydrochloride, also known as Loprinone Hydrochloride, is a potent phosphodiesterase (PDE) 3 inhibitor with an IC50 of 0.35 μM. It is utilized in research for heart failure due to its positive inotropic and vasodilative effects, and it also demonstrates anti-inflammatory activity.
  • Potent PDE3 inhibitor (IC50 of 0.35 μM)
  • Used in research for heart failure
  • Exhibits positive inotropic and vasodilative effects
  • Demonstrates anti-inflammatory activity
  • Modulates inflammation associated with myocardial ischemia-reperfusion injury in rats
  • Reduces histological evidence of myocardial injury
  • Decreases pro-inflammatory cytokines (TNF-α and IL-1β)
  • Reduces adhesion molecules (ICAM-1 and P-Selectin)
  • Decreases nitrotyrosine formation and NF-κB expression
  • Reduces PAR formation and apoptosis