Learn More
Medchemexpress LLC Olprinone hydrochloride | 119615-63-3 | 99.9% | C14H11ClN4O | 50 MG
SDP
Catalog No. 5000353566
Encompass_Preferred
Olprinone Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, showing IC50s of 0.35 μM for PDE3. It is utilized in research for heart failure due to its positive inotropic and vasodilative effects, and also demonstrates anti-inflammatory activity.
  • Potent PDE3 inhibitor
  • Used in research for heart failure
  • Exhibits positive inotropic and vasodilative effects
  • Demonstrates anti-inflammatory activity
  • Modulates inflammation associated with myocardial ischemia-reperfusion injury
  • Reduces pro-inflammatory cytokines such as TNF-α and IL-1β
  • Decreases expression of adhesion molecules like ICAM-1 and P-Selectin
  • Inhibits apoptosis related to myocardial injury
Catalog No. 50-003-53566 Supplier Medchemexpress LLC Supplier No. HY1425450MG
Olprinone Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, showing IC50s of 0.35 μM for PDE3. It is utilized in research for heart failure due to its positive inotropic and vasodilative effects, and also demonstrates anti-inflammatory activity.
  • Potent PDE3 inhibitor
  • Used in research for heart failure
  • Exhibits positive inotropic and vasodilative effects
  • Demonstrates anti-inflammatory activity
  • Modulates inflammation associated with myocardial ischemia-reperfusion injury
  • Reduces pro-inflammatory cytokines such as TNF-α and IL-1β
  • Decreases expression of adhesion molecules like ICAM-1 and P-Selectin
  • Inhibits apoptosis related to myocardial injury