Learn More
Medchemexpress LLC Olprinone hydrochloride | 119615-63-3 | MFCD01749474 | ≥98.0% | 286.72 g·mol⁻¹ | C14H11ClN4O | 5 MG
SDP
Catalog No. 5000221491
Encompass_Preferred
Olprinone hydrochloride is a research-grade phosphodiesterase 3 (PDE3) inhibitor provided as an analytical standard for pharmacological and biochemical studies, commonly used in investigations of cardiac inotropy and vasodilation. The material is characterized for laboratory use and supplied in small pack sizes for precise dosing in experimental workflows.
  • Research-grade analytical standard suitable for assay development and characterization.
  • Selective phosphodiesterase 3 (PDE3) inhibitory activity relevant to cardiac function studies.
  • Provided in small pack sizes for precise dosing and handling in the laboratory.
  • Molecular formula C14H11ClN4O and molecular weight 286.72 g·mol⁻¹ for reproducible calculations.
  • Available with high purity specifications to support reliable experimental results.
Catalog No. 50-002-21491 Supplier Medchemexpress LLC Supplier No. HY142545MG
Olprinone hydrochloride is a research-grade phosphodiesterase 3 (PDE3) inhibitor provided as an analytical standard for pharmacological and biochemical studies, commonly used in investigations of cardiac inotropy and vasodilation. The material is characterized for laboratory use and supplied in small pack sizes for precise dosing in experimental workflows.
  • Research-grade analytical standard suitable for assay development and characterization.
  • Selective phosphodiesterase 3 (PDE3) inhibitory activity relevant to cardiac function studies.
  • Provided in small pack sizes for precise dosing and handling in the laboratory.
  • Molecular formula C14H11ClN4O and molecular weight 286.72 g·mol⁻¹ for reproducible calculations.
  • Available with high purity specifications to support reliable experimental results.