Learn More
Selleck Chemical LLC Olprinone Hydrochloride E4979-100MG
SDP
Catalog No. 5000041631
Encompass_Preferred
Olprinone hydrochloride(Loprinone Hydrochloride E-1020) is a potent inhibitor of phosphodiesterase (PDE) 3 with an IC50 value of 0 35 M for PDE3 respectively Olprinone Hydrochloride is used in the research of heart failure treatment due to its positive inotropic and vasodilatory effects It also exhibits anti-inflammatory properties
Catalog No. 50-000-41631 Supplier Selleck Chemical LLC Supplier No. E4979100MG
Olprinone hydrochloride(Loprinone Hydrochloride E-1020) is a potent inhibitor of phosphodiesterase (PDE) 3 with an IC50 value of 0 35 M for PDE3 respectively Olprinone Hydrochloride is used in the research of heart failure treatment due to its positive inotropic and vasodilatory effects It also exhibits anti-inflammatory properties