Learn More
Medchemexpress LLC Olsalazine Disodium | 6054-98-4 | 99.8% | 1 G
SDP
Catalog No. 5000406313
Encompass_Preferred
Olsalazine Disodium is an orally active prodrug of 5-ASA. It can inhibit cell proliferation and induce apoptosis, and reduce DAI and MPO activity and inhibit inflammatory cytokine levels. It is used in research for cancer, inflammation, and metabolic diseases, including colorectal cancer, inflammatory bowel disease (IBD), and hyperuricemia.
  • Inhibits cell proliferation
  • Induces apoptosis
  • Reduces DAI and MPO activity
  • Inhibits inflammatory cytokine levels
  • Useful for research on colorectal cancer
  • Useful for research on inflammatory bowel disease (IBD)
  • Useful for research on hyperuricemia
Catalog No. 50-004-06313 Supplier Medchemexpress LLC Supplier No. HYB01741G
Olsalazine Disodium is an orally active prodrug of 5-ASA. It can inhibit cell proliferation and induce apoptosis, and reduce DAI and MPO activity and inhibit inflammatory cytokine levels. It is used in research for cancer, inflammation, and metabolic diseases, including colorectal cancer, inflammatory bowel disease (IBD), and hyperuricemia.
  • Inhibits cell proliferation
  • Induces apoptosis
  • Reduces DAI and MPO activity
  • Inhibits inflammatory cytokine levels
  • Useful for research on colorectal cancer
  • Useful for research on inflammatory bowel disease (IBD)
  • Useful for research on hyperuricemia