Learn More
Medchemexpress LLC Olsalazine (disodium) | 6054-98-4 | 99.8% | 10 G
SDP
Catalog No. 5000405394
Encompass_Preferred
Olsalazine Disodium is an orally active prodrug of 5-ASA. It is utilized in research for cancer, inflammation, and metabolic diseases, including colorectal cancer, inflammatory bowel disease (IBD), and hyperuricemia. This compound can inhibit cell proliferation, induce apoptosis, reduce DAI and MPO activity, and inhibit inflammatory cytokine levels.
  • Orally active prodrug of 5-ASA
  • Inhibits cell proliferation
  • Induces apoptosis
  • Reduces DAI and MPO activity
  • Inhibits inflammatory cytokine levels
  • Suitable for research in cancer
  • Suitable for research in inflammation
  • Suitable for research in metabolic diseases
Catalog No. 50-004-05394 Supplier Medchemexpress LLC Supplier No. HYB017410G
Add to Cart
Add to Cart
Olsalazine Disodium is an orally active prodrug of 5-ASA. It is utilized in research for cancer, inflammation, and metabolic diseases, including colorectal cancer, inflammatory bowel disease (IBD), and hyperuricemia. This compound can inhibit cell proliferation, induce apoptosis, reduce DAI and MPO activity, and inhibit inflammatory cytokine levels.
  • Orally active prodrug of 5-ASA
  • Inhibits cell proliferation
  • Induces apoptosis
  • Reduces DAI and MPO activity
  • Inhibits inflammatory cytokine levels
  • Suitable for research in cancer
  • Suitable for research in inflammation
  • Suitable for research in metabolic diseases