Learn More
Apexbio Technology LLC Olsalazine Sodium 6054-98-4 10mM (in 1mL H2O)
SDP
Catalog No. 5000095305
Encompass_Preferred
Olsalazine Sodium (CAS 6054-98-4) is a small-molecule anti-inflammatory agent It is designed to modulate inflammatory pathways thereby reducing the production of local inflammatory mediators Olsalazine Sodium exerts its biological activity primarily through inhibition of prostaglandin and leukotriene synthesis pathways after biotransformation by gut microbiota into 5-aminosalicylic acid (5-ASA) In experimental settings researchers employ olsalazine sodium to investigate disease mechanisms underlying colonic inflammation and to evaluate therapeutic responses in models of intestinal inflammation Currently precise IC50 values for olsalazine sodium have not been reported in the literature Based on these pharmacological properties olsalazine sodium holds research potential in inflammatory bowel disease (IBD) especially ulcerative colitis
Catalog No. 50-000-95305 Supplier Apexbio Technology LLC Supplier No. A84905.1
Olsalazine Sodium (CAS 6054-98-4) is a small-molecule anti-inflammatory agent It is designed to modulate inflammatory pathways thereby reducing the production of local inflammatory mediators Olsalazine Sodium exerts its biological activity primarily through inhibition of prostaglandin and leukotriene synthesis pathways after biotransformation by gut microbiota into 5-aminosalicylic acid (5-ASA) In experimental settings researchers employ olsalazine sodium to investigate disease mechanisms underlying colonic inflammation and to evaluate therapeutic responses in models of intestinal inflammation Currently precise IC50 values for olsalazine sodium have not been reported in the literature Based on these pharmacological properties olsalazine sodium holds research potential in inflammatory bowel disease (IBD) especially ulcerative colitis