Learn More
Stoelting™ Olsen-Hegar Needle Holder
SDP
Catalog No. 10001989 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
4.5 in.
4.75 in.
5.5 in.
6.5 in.
6.75 in.
Model:
Extra delicate, Serrated
Extra delicate, Smooth
N/A
Tungsten carbide
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-989 4.5 in. Tungsten carbide
10-001-987 4.75 in. Extra delicate, Serrated
10-001-869 4.75 in. Extra delicate, Smooth
10-001-988 5.5 in. N/A
10-001-990 5.5 in. Tungsten carbide
10-001-871 6.5 in. N/A
10-001-991 6.75 in. Tungsten carbide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-001-989 Supplier Stoelting™ Supplier No. 5218192P
Add to Cart
Add to Cart
Stoelting™ Olsen-Hegar Needle Holder are designed to give precise control for various needle types. Features include: Stainless steel
Specifications
For Use With (Application) Neuroscience
Length (English) 4.5 in.
Model Tungsten carbide