Learn More
Sklar Instruments Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Delicate, Finger Rings, Stainless Steel. EA/1.
SDP
Supplier:  Sklar Instruments 202747
Encompass_Preferred
Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Delicate, Finger Rings, Stainless Steel. Sklar's Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. This product is straight with delicate, cross-serrated tips, and an overall length of 4-3/4 inches.
Catalog No. 50-143-7485