Learn More
Fine Science Tools Olsen-Hegar Needle Holders with Suture Cutters (Left-Handed), 12 cm
SDP
Catalog No. NC1211817
Encompass
Product: Olsen-Hegar Needle Holders with Suture Cutters (Left-Handed)/12cm/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 7.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Unit: Each
Catalog No. NC1211817 Supplier Fine Science Tools Supplier No. 1210212
Product: Olsen-Hegar Needle Holders with Suture Cutters (Left-Handed)/12cm/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 7.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Unit: Each