Learn More
Sklar Instruments TC Olsen-Hegar Needle Holder, Sklar 5-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184179 Shop All Sklar Instruments Products
Encompass_Preferred
TC Olsen-Hegar Needle Holder, Sklar. 5-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. The Surgi-OR Olsen-Hegar TC Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips, and an overall length of 5-1/2 inches.
Catalog No. 50-118-4179 Supplier Sklar Instruments Supplier No. 95147
TC Olsen-Hegar Needle Holder, Sklar. 5-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. The Surgi-OR Olsen-Hegar TC Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips, and an overall length of 5-1/2 inches.