Learn More
Medchemexpress LLC HY-17641 5mg Medchemexpress, Olumacostat glasaretil CAS:1261491-89-7 Purity:>98%
SDP
Catalog No. 502082006
Encompass_Preferred
Medchemexpress, HY-17641 5mg Olumacostat glasaretil CAS:1261491-89-7 Olumacostat glasaretil is a small molecule inhibitor of acetyl coenzyme A carboxylase (ACC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2006 Supplier Medchemexpress LLC Supplier No. HY176415MG
Medchemexpress, HY-17641 5mg Olumacostat glasaretil CAS:1261491-89-7 Olumacostat glasaretil is a small molecule inhibitor of acetyl coenzyme A carboxylase (ACC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.