Learn More
Cayman Chemical olyl TrIfluoromthyl ketn 10mg
SDP
Catalog No. 502892883
Encompass_Preferred
An analog of oleic acid in which the COOH group is replaced by trifluoromethyl ketone; it’s a potent inhibitor of FAAH, in both human and rat; in transfected COS-7 cells, 10 µM OTK inhibits 95.7% of human FAAH activity and 94.8% of rat FAAH activity
Catalog No. 50-289-2883 Supplier Cayman Chemical Supplier No. 6264010 MG
An analog of oleic acid in which the COOH group is replaced by trifluoromethyl ketone; it’s a potent inhibitor of FAAH, in both human and rat; in transfected COS-7 cells, 10 µM OTK inhibits 95.7% of human FAAH activity and 94.8% of rat FAAH activity