Learn More
Cayman Chemical olyl TrIfluoromthyl ketn 50mg
SDP
Catalog No. 502893065
Encompass_Preferred
An analog of oleic acid in which the COOH group is replaced by trifluoromethyl ketone; it’s a potent inhibitor of FAAH, in both human and rat; in transfected COS-7 cells, 10 µM OTK inhibits 95.7% of human FAAH activity and 94.8% of rat FAAH activity
Catalog No. 50-289-3065 Supplier Cayman Chemical Supplier No. 6264050 MG
An analog of oleic acid in which the COOH group is replaced by trifluoromethyl ketone; it’s a potent inhibitor of FAAH, in both human and rat; in transfected COS-7 cells, 10 µM OTK inhibits 95.7% of human FAAH activity and 94.8% of rat FAAH activity