Learn More
Cayman Chemical Oleyloxyethyl Phosphorylcholine, Purity: ≥98%, CAS: 84601-19-4, 10 mg
SDP
Catalog No. 502892227
Encompass_Preferred
Oleyloxyethyl phosphorylcholine is an inhibitor of PLA2 with an IC50 value of 6.2 µM for porcine pancreatic PLA2.{879}
Catalog No. 50-289-2227 Supplier Cayman Chemical Supplier No. 7056010 MG
Oleyloxyethyl phosphorylcholine is an inhibitor of PLA2 with an IC50 value of 6.2 µM for porcine pancreatic PLA2.{879}