Learn More
Genesee Scientific Corporation Olympus 0.6ml Microtubes, Natural, 1000/pk
SDP
Catalog No. NC1770723
Encompass
Certified free from RNase, DNase, DNA & PCR inhibitors. Also Endotoxin-FreeAutoclavable, Temp Range: -90ºC to 121ºCRated to >20,000 x gGraduations every 100µl - easier estimation of contents!Frosted side writing surfaceExceptionally clear - glass-like clarityMade in the USA
Catalog No. NC1770723 Supplier Genesee Scientific Corporation Supplier No. 24272
Add to Cart
Add to Cart
Certified free from RNase, DNase, DNA & PCR inhibitors. Also Endotoxin-FreeAutoclavable, Temp Range: -90ºC to 121ºCRated to >20,000 x gGraduations every 100µl - easier estimation of contents!Frosted side writing surfaceExceptionally clear - glass-like clarityMade in the USA