Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Diagnostics Olympus™ Electrodes

Catalog No. 22293263 Shop All Phoenix Diagnostics, Inc Products
Change view
Click to view available options
Type:
Cl- Electrode
K+ Electrode
Reference Electrode
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-293-263 Cl- Electrode
22-293-262 Reference Electrode
22-293-261 K+ Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-293-263 Supplier Phoenix Diagnostics Supplier No. E920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reference Electrodes for Olympus AU Series analyzers.

  • Replacement electrodes available for use in all Olympus AU series analyzers
  • Na+, K+, Cl-, and Reference electrodes available

Specifications

Type Cl- Electrode
Sample Type Whole Blood, Plasma, Serum, Urine
For Use With (Equipment) Olympus™ AU series analyzer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.