Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OM-153 | 2406278-81-5 | 99.3% | C28H24FN7O2 | 1 ML
SDP

Catalog No. 5000355210
Encompass_Preferred

OM-153 is a potent, orally active tankyrase inhibitor targeting TNKS1/2 with high specificity. It demonstrates strong inhibitory activity against Wnt/β-catenin signaling, which is crucial for cellular proliferation, and has shown antiproliferative effects in COLO 320DM cells.

  • Potently inhibits TNKS1 (IC50: 13 nM) and TNKS2 (IC50: 2 nM).
  • Effectively inhibits Wnt/β-catenin signaling (IC50: 0.63 nM).
  • Exhibits antiproliferative activity in COLO 320DM cells (GI50: 10 nM).
  • Inhibits WNT/β-catenin, YAP, and MYC signaling pathways.
  • Reduces WNT/β-catenin signaling and tumor progression in COLO 320DM xenografts.
  • Potentiates anti-PD-1 immune checkpoint inhibition in melanoma models.

Catalog No. 50-003-55210 Supplier Medchemexpress LLC Supplier No. HY14526710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OM-153 is a potent, orally active tankyrase inhibitor targeting TNKS1/2 with high specificity. It demonstrates strong inhibitory activity against Wnt/β-catenin signaling, which is crucial for cellular proliferation, and has shown antiproliferative effects in COLO 320DM cells.

  • Potently inhibits TNKS1 (IC50: 13 nM) and TNKS2 (IC50: 2 nM).
  • Effectively inhibits Wnt/β-catenin signaling (IC50: 0.63 nM).
  • Exhibits antiproliferative activity in COLO 320DM cells (GI50: 10 nM).
  • Inhibits WNT/β-catenin, YAP, and MYC signaling pathways.
  • Reduces WNT/β-catenin signaling and tumor progression in COLO 320DM xenografts.
  • Potentiates anti-PD-1 immune checkpoint inhibition in melanoma models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.