Learn More
Aviva Systems Biology Corp OMA1 Antibody - middle region (ARP52818_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52818_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OMA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OMA1. Alternate names: 2010001O09Rik, DAB1, FLJ33782, MPRP-1, YKR087C, ZMPOMA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-771