Learn More
TARGETMOL CHEMICALS INC OMADACYCLINE 5MG
SDP
Catalog No. 502576700
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Omadacycline (Amadacycline) a new tetracycline antibiotic can inhibit the 30s subunit of bacterial ribosome. purity: 99%
Catalog No. 50-257-6700 Supplier TARGETMOL CHEMICALS INC Supplier No. T51895MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Omadacycline (Amadacycline) a new tetracycline antibiotic can inhibit the 30s subunit of bacterial ribosome. purity: 99%