Learn More
Medchemexpress LLC Omadacycline-d9 | 2272886-41-4 | 99.21% | 565.71 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY14865S500UG
Encompass_Preferred
Omadacycline-d9 (PTK 0796-d9; Amadacycline-d9) is the deuterium labeled Omadacycline (HY-14865) that can be used as the internal standard for the analysis of Omadacycline.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62091