Learn More
Medchemexpress LLC Omadacycline-d9 (PTK 0796-d9; Amadacycline-d9) | 2272886-41-4 | 99.2% | 565.71 | 1 MG
SDP
Catalog No. 5000360986
Encompass_Preferred
Omadacycline-d9 (PTK 0796-d9; Amadacycline-d9) is the deuterium labeled Omadacycline that can be used as the internal standard for the analysis of Omadacycline. This product is for research use only.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60986 Supplier Medchemexpress LLC Supplier No. HY14865S1MG
Omadacycline-d9 (PTK 0796-d9; Amadacycline-d9) is the deuterium labeled Omadacycline that can be used as the internal standard for the analysis of Omadacycline. This product is for research use only.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS