Learn More
Medchemexpress LLC HY-14865C 10mg Medchemexpress, Omadacycline (hydrochloride) CAS:1196800-39-1 Purity:>98%
SDP
Catalog No. 502029073
Encompass_Preferred
Medchemexpress, HY-14865C 10mg Omadacycline (hydrochloride) CAS:1196800-39-1 Omadacycline hydrochloride is novel, aminomethyl tetracycline antibiotic being developed for the treatment of community-acquired bacterial infections. The ED50 for Escherichia coli is 2.02 mg/kg. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9073 Supplier Medchemexpress LLC Supplier No. HY14865C10MG
Medchemexpress, HY-14865C 10mg Omadacycline (hydrochloride) CAS:1196800-39-1 Omadacycline hydrochloride is novel, aminomethyl tetracycline antibiotic being developed for the treatment of community-acquired bacterial infections. The ED50 for Escherichia coli is 2.02 mg/kg. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.