Learn More
Selleck Chemical LLC Omadacycline hydrochloride E4983-1G
SDP
Catalog No. 5000041159
Encompass_Preferred
Omadacycline hydrochloride (PTK0796 hydrochloride) is a first-in-class orally active member of the tetracycline class of antibiotics Omadacycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit Omadacycline hydrochloride exhibits broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria as well as atypical bacteria
Catalog No. 50-000-41159 Supplier Selleck Chemical LLC Supplier No. E49831G
Omadacycline hydrochloride (PTK0796 hydrochloride) is a first-in-class orally active member of the tetracycline class of antibiotics Omadacycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit Omadacycline hydrochloride exhibits broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria as well as atypical bacteria